Aluminium fluoride, anhydrous 99,9%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Aluminum fluoride
IN1015-10G
IN1015-25G
APOSIN1015-10GEA
7080
CZK
APOSIN1015-10G
APOSIN1015-25G
Aluminium fluoride, anhydrous 99,9%
Aluminium fluoride
Vzorec:
AlF₃ MW: 83.98 Bod tání: 1291 °C (subl.) Hustota: 3.1 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003426 CAS číslo: 7784-18-1 EINECS: 232-051-1 Merck Index: 12,00351 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...