Tisk…

Hexaamminecobalt trichloride

Dodavatel: Apollo Scientific
Varování

Synonyms: Hexaamminecobalt(iii)chloride, Hexammine cobalt(III) chloride, Hexaamminecobalt (III) chloride

IN0745-25G IN0745-100G IN0745-500G
APOSIN0745-25GEA 3540 CZK
APOSIN0745-25G APOSIN0745-100G APOSIN0745-500G
Hexaamminecobalt trichloride
Hexaamminecobalt trichloride
Vzorec: H₁₈CoCl₃N₆
MW: 267,47 g/mol
Bod tání: 217 °C
Hustota: 1,71 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036304
CAS číslo: 10534-89-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR