Zeatin riboside
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
trans-Zeatinriboside, trans-Zeatin-riboside
BIZ0937-100MG
APOSBIZ0937-100MGEA
4370
CZK
APOSBIZ0937-100MG
Zeatin riboside
Zeatin riboside
Used for plant regulation from tomato, Brassica nigra and vigna sublobata protoplasts.
Vzorec:
C₁₅H₂₁N₅O₅ MW: 351,363 g/mol Bod tání: 176…180 °C Storage Temperature: Freezer |
MDL Number:
MFCD00036809 CAS číslo: 6025-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...