trans-Zeatin
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-(4-Hydroxy-3-methyl-2-butenylamino)purine
BIZ0917-100MG
BIZ0917-250MG
BIZ0917-50MG
APOSBIZ0917-100MGEA
3740
CZK
APOSBIZ0917-100MG
APOSBIZ0917-250MG
APOSBIZ0917-50MG
trans-Zeatin
trans-Zeatin
Cytokinin growth regulator.
Vzorec:
C₁₀H₁₃N₅O MW: 219,25 g/mol Bod tání: 208…210 °C Storage Temperature: Freezer |
MDL Number:
MFCD00213654 CAS číslo: 1637-39-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...