Tisk…

Withaferin A

Dodavatel: Apollo Scientific
Varování

Synonyms: 5,6-Epoxy-4,27-dihydroxy-1-oxowitha-2,24-dienolide

BIW4202-10MG BIW4202-5MG
APOSBIW4202-10MGEA 12400 CZK
APOSBIW4202-10MG APOSBIW4202-5MG
Withaferin A
Withaferin A

Possible carcinogen.

Vzorec: C₂₈H₃₈O₆
MW: 470,6 g/mol
Bod tání: 252…253 °C
Storage Temperature: Freezer
CAS číslo: 5119-48-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR