Tisk…

Verapamil hydrochloride

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 5-((3,4-Dimethoxyphenethyl)(methyl)amino)-2-(3,4-dimethoxyphenyl)-2-isopropylpentanenitrile hydrochloride, Verapamil HCl, 5-[(3,4-Dimethoxyphenethyl)methylamino]-2-(3,4-dimethoxyphenyl)-2-isopropylvaleronitrile Hydrochloride, (±)-Verapamil hydrochloride

BIV1326-25G BIV1326-5G
APOSBIV1326-25GEA 3810 CZK
APOSBIV1326-25G APOSBIV1326-5G
Verapamil hydrochloride
Verapamil hydrochloride

Calcium channel blocker.

Vzorec: C₂₇H₃₈N₂O₄
MW: 491,07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00055208
CAS číslo: 152-11-4
EINECS: 205-800-5
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR