Tisk…

Verruculogen (from Penicillium verruculosum)

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: NA 209A

BIV1002-10MG BIV1002-1MG BIV1002-5MG
APOSBIV1002-10MGEA 29590 CZK
APOSBIV1002-10MG APOSBIV1002-1MG APOSBIV1002-5MG
Verruculogen (from Penicillium verruculosum)
Verruculogen
Vzorec: C₂₇H₃₃N₃O₇
Bod tání: 230…233 °C
Storage Temperature: Freezer
CAS číslo: 12771-72-1
UN: 2811
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR