Tisk…

U0126 ≥99%

Dodavatel: Apollo Scientific
Varování

BIU1095-10MG BIU1095-25MG BIU1095-5MG
APOSBIU1095-10MGEA 8270 CZK
APOSBIU1095-10MG APOSBIU1095-25MG APOSBIU1095-5MG
U0126 ≥99%
U0126

Selective inhibitor of the mitogen-activated protein kinase kinases, MEK-1 and MEK-2, with 100-fold higher potency than PD 98059, but is, at most, a weak inhibitor of PKC, Raf, ERK, JNK, MEKK, MKK-3, MKK-4/SEK, MKK-6, Abl, Cdk2 and Cdk4. U0126 is an inhibitor of AP-1 transactivation in cell-based reporter assays. J. Biol. Chem. 273: 18623-18632 (1998).

Vzorec: C₁₈H₁₆N₆S₂
MW: 380.49 g/mol
Bod tání: 172…173 °C
Storage Temperature: Freezer
MDL Number: MFCD01861186
CAS číslo: 109511-58-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR