Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) dihydrate
Dodavatel: Apollo Scientific
|
Varování
|
BIU0402-1G
BIU0402-500MG
APOSBIU0402-1GEA
1990
CZK
APOSBIU0402-1G
APOSBIU0402-500MG
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) dihydrate
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) dihydrate
Vzorec:
C₉H₁₂N₂Na₃O₁₅P₃·2H₂O Bod tání: >140 °C Storage Temperature: Freezer |
CAS číslo:
116295-90-0 EINECS: 243-347-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...