Thapsigargin
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
(3S,3aR,4S,6S,6aR,7S,8S,9bS)-6-Acetoxy-4-(butyryloxy)-3,3a-dihydroxy-3,6,9-trimethyl-8-(((Z)-2-methylbut-2-enoyl)oxy)-2-oxo-2,3,3a,4,5,6,6a,7,8,9b-decahydroazuleno[4,5-b]furan-7-yl octanoate
BIT4520-10MG
BIT4520-1MG
BIT4520-5MG
APOSBIT4520-10MGEA
24170
CZK
APOSBIT4520-10MG
APOSBIT4520-1MG
APOSBIT4520-5MG
Thapsigargin
Thapsigargin
Potent calcium releaser
Vzorec:
C₃₄H₅₀O₁₂ MW: 650,762 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00083511 CAS číslo: 67526-95-8 UN: 2811 ADR: 6.1,III |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...