Tisk…

L-Tyrosine disodium salt

Dodavatel: Apollo Scientific
Varování

BIT3156-100G BIT3156-25G
APOSBIT3156-100GEA 2690 CZK
APOSBIT3156-100G APOSBIT3156-25G
L-Tyrosine disodium salt
L-Tyrosine disodium salt
Vzorec: C₁₈H₂₀N₂Na₂O₆
MW: 406.346 g/mol
Storage Temperature: Refrigerator
CAS číslo: 69847-45-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR