Tisk…

Tetrazólium modř chlorid

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Blue Tetrazolium, 3,3′-(3,3′-Dimethoxy[1,1′-biphenyl]-4,4′-diyl)-bis(2,5-diphenyl-2H-tetrazolium) dichloride, 3,3'-(3,3'-Dimethoxy-4,4'-biphenylene)bis(2,5-diphenyl-2H-tetrazolium chloride), BTC, 3,3′-Dianisole-4,4′-bis(3,5-diphenyltetrazolium chloride), Blue Tetrazolium chloride

BIT3025-10G BIT3025-5G BIT3025-25G
APOSBIT3025-10GEA 2690 CZK
APOSBIT3025-10G APOSBIT3025-5G APOSBIT3025-25G
Tetrazólium modř chlorid
Tetrazólium modř chlorid
Vzorec: C₄₀H₃₂Cl₂N₈O₂
Storage Temperature: Ambient
MDL Number: MFCD00040933
CAS číslo: 1871-22-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR