Tisk…

Tyrphostin AG 1478 free base 99+%

Dodavatel: Apollo Scientific
Varování

BIT1101-100MG BIT1101-25MG
APOSBIT1101-100MGEA 12200 CZK
APOSBIT1101-100MG APOSBIT1101-25MG
Tyrphostin AG 1478 free base 99+%
Tyrphostin AG 1478

Very potent EGF receptor kinase (tyrosine kinase) inhibitor (IC50 = 3 nM). Very weak on PDGF receptor and HER2-NEU kinases (IC50 >100 uM). Eur. J. Biochem. 225: 1047-1053 (1994).

Vzorec: C₁₆H₁₄ClN₃O₂
Storage Temperature: Freezer
MDL Number: MFCD00270914
CAS číslo: 153436-53-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR