Tisk…

Triacsin C (from Streptomyces species)

Dodavatel: Apollo Scientific
Varování

BIT1004-1MG BIT1004-100UG
APOSBIT1004-1MGEA 31840 CZK
APOSBIT1004-1MG APOSBIT1004-100UG
Triacsin C (from Streptomyces species)
Triacsin C

Fungal metabolite.

Vzorec: C₁₁H₁₇N₃O
Storage Temperature: Freezer
CAS číslo: 76896-80-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR