Tisk…

Ticaracillin disodium salt

Dodavatel: Apollo Scientific
Varování

Synonyms: Ticarcillin disodium salt

BIT0186-1G BIT0186-500MG BIT0186-5G
APOSBIT0186-1GEA 2320 CZK
APOSBIT0186-1G APOSBIT0186-500MG APOSBIT0186-5G
Ticaracillin disodium salt
Ticaracillin disodium salt

Inhibits bacterial cell wall synthesis. Highly active against gram negative bacteria such as Agrobacterium strains

Vzorec: C₁₅H₁₄N₂Na₂O₆S₂
Storage Temperature: Refrigerator
MDL Number: MFCD07787410
CAS číslo: 4697-14-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR