Tisk…

3,4-Di-O-acetyl-6-deoxy-L-glucal

Dodavatel: Apollo Scientific

Synonyms: 3,4-Di-O-acetyl-1,5-anhydro-2,6-dideoxy-L-arabino-hex-1-enitol, 3,4-Di-O-acetyl-L-rhamnal

BISY007-1G BISY007-5G
APOSBISY007-1GEA 2690 CZK
APOSBISY007-1G APOSBISY007-5G
3,4-Di-O-acetyl-6-deoxy-L-glucal
3,4-Di-O-acetyl-6-deoxy-L-glucal
Vzorec: C₁₀H₁₄O₅
Storage Temperature: Ambient
MDL Number: MFCD00074970
CAS číslo: 34819-86-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR