Tisk…

Camostat methanesulphate

Dodavatel: Apollo Scientific

BISN0316-250MG BISN0316-500MG BISN0316-1G
APOSBISN0316-250MGEA 3110 CZK
APOSBISN0316-250MG APOSBISN0316-500MG APOSBISN0316-1G
Camostat methanesulphate
Camostat methanesulphate

Camostat mesylateis a trypsin-like protease inhibitor, inhibits airway epithelial sodium channel (ENaC) function with IC50 of 50 nM, less potent to trypsin, prostasin and matriptase. This inhibitor has shown to block TMPRSS2 which facilitates Coronavirus entry into host cell.

Vzorec: C₂₀H₂₂N₄O₅·CH₃SO₃H
Storage Temperature: Refrigerator
MDL Number: MFCD00941410
CAS číslo: 59721-29-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR