Tisk…

CCT-196969

Dodavatel: Apollo Scientific

BISN0303-1MG BISN0303-5MG BISN0303-10MG
APOSBISN0303-1MGEA 1870 CZK
APOSBISN0303-1MG APOSBISN0303-5MG APOSBISN0303-10MG
CCT-196969
CCT-196969

MAPK/ERK Signalling Pathway

Vzorec: C₂₇H₂₄FN₇O₃
MW: 513,50 g/mol
Storage Temperature: Ambient
CAS číslo: 1163719-56-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR