SB242235
Dodavatel: Apollo Scientific
Synonyms:
4-(4-(4-Fluorophenyl)-1-(piperidin-4-yl)-1H-imidazol-5-yl)-2-methoxypyrimidine, SB 242235, 4-[5-(4-fluorophenyl)-3-piperidin-4-ylimidazol-4-yl]-2-methoxypyrimidine
BISN0286-1MG
BISN0286-5MG
BISN0286-25MG
APOSBISN0286-1MGEA
2480
CZK
APOSBISN0286-1MG
APOSBISN0286-5MG
APOSBISN0286-25MG
SB242235
SB242235
MAPK/ERK Signalling Pathway
Vzorec:
C₁₉H₂₀FN₅O MW: 353,4 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08690575 CAS číslo: 193746-75-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...