Tisk…

Pluripotin

Dodavatel: Apollo Scientific

Synonyms: N-(3-(7-((1,3-Dimethyl-1H-pyrazol-5-yl)amino)-1-methyl-2-oxo-1,2-dihydropyrimido[4,5-d]pyrimidin-3(4H)-yl)-4-methylphenyl)-3-(trifluoromethyl)benzamide

BISN0279-5MG BISN0279-25MG
APOSBISN0279-5MGEA 6190 CZK
APOSBISN0279-5MG APOSBISN0279-25MG
Pluripotin
Pluripotin

MAPK/ERK Signalling Pathway

Vzorec: C₂₇H₂₅F₃N₈O₂
MW: 550,546 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20527253
CAS číslo: 839707-37-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR