PH-797804
Dodavatel: Apollo Scientific
Synonyms:
3-(4-(2,4-difluorobenzyloxy)-3-bromo-6-methyl-2-oxopyridin-1(2H)-yl)-N,4-dimethylbenzamide, 3-(3-Bromo-4-((2,4-difluorobenzyl)oxy)-6-methyl-2-oxopyridin-1(2H)-yl)-N,4-dimethylbenzamide, PH797804, PH 797804, 3-[3-bromo-4-[(2,4-difluorophenyl)methoxy]-6-methyl-2-oxopyridin-1-yl]-N,4-dimethylbenzamide, PH-797804
BISN0276-25MG
APOSBISN0276-25MGEA
5060
CZK
APOSBISN0276-25MG
PH-797804
PH-797804
MAPK/ERK Signalling Pathway
Vzorec:
C₂₂H₁₉BrF₂N₂O₃ MW: 477,3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18251426 CAS číslo: 586379-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...