Skepinone-L
Dodavatel: Apollo Scientific
Synonyms:
(R)-2-((2,4-Difluorophenyl)amino)-7-(2,3-dihydroxypropoxy)-10,11-dihydro-5H-dibenzo[a,d][7]annulen-5-one, 2-[(2,4-difluorophenyl)amino]-7-[(2R)-2,3-dihydroxypropoxy]-10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5-one
BISN0248-1MG
BISN0248-5MG
BISN0248-10MG
APOSBISN0248-1MGEA
2940
CZK
APOSBISN0248-1MG
APOSBISN0248-5MG
APOSBISN0248-10MG
Skepinone-L
Skepinone-L
MAPK/ERK Signalling Pathway
Vzorec:
C₂₄H₂₁F₂NO₄ MW: 425,40 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22572738 CAS číslo: 1221485-83-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...