CC-90003
Dodavatel: Apollo Scientific
Synonyms:
CC90003, N-(2-((2-((2-methoxy-5-methylpyridin-4-yl)amino)-5-(trifluoromethyl)pyrimidin-4-yl)amino)-5-methylphenyl)acrylamide
BISN0236-25MG
BISN0236-50MG
APOSBISN0236-25MGEA
6220
CZK
APOSBISN0236-25MG
APOSBISN0236-50MG
CC-90003
CC-90003
MAPK/ERK Signalling Pathway
Vzorec:
C₂₂H₂₁F₃N₆O₂ MW: 458,44 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31692404 CAS číslo: 1621999-82-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...