Tisk…

Raf1 Kinase Inhibitor I

Dodavatel: Apollo Scientific

BISN0227-1MG BISN0227-5MG BISN0227-10MG
APOSBISN0227-1MGEA 622 CZK
APOSBISN0227-1MG APOSBISN0227-5MG APOSBISN0227-10MG
Raf1 Kinase Inhibitor I
Raf1 Kinase Inhibitor I

MAPK/ERK Signalling Pathway

Vzorec: C₁₅H₈Br₂INO₂
Storage Temperature: Ambient
CAS číslo: 220904-83-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR