Tisk…

ZM-336372

Dodavatel: Apollo Scientific

Synonyms: 3-(Dimethylamino)-N-(3-(4-hydroxybenzamido)-4-methylphenyl)benzamide, ZM 336372

BISN0205-5MG
APOSBISN0205-5MGEA 1370 CZK
APOSBISN0205-5MG
ZM-336372
ZM-336372

MAPK/ERK Signalling Pathway

Vzorec: C₂₃H₂₃N₃O₃
MW: 389,455 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02683971
CAS číslo: 208260-29-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR