Tisk…

DEL-22379

Dodavatel: Apollo Scientific

Synonyms: DEL 22379, N-(3-((5-Methoxy-1H-indol-3-yl)methylene)-2-oxoindolin-5-yl)-3-(piperidin-1-yl)propanamide, DEL22379, N-[(3Z)-3-[(5-methoxy-1H-indol-3-yl)methylidene]-2-oxo-1H-indol-5-yl]-3-piperidin-1-ylpropanamide, DEL-22379

BISN0204-2MG BISN0204-5MG BISN0204-10MG
APOSBISN0204-2MGEA 4960 CZK
APOSBISN0204-2MG APOSBISN0204-5MG APOSBISN0204-10MG
DEL-22379
DEL-22379

MAPK/ERK Signalling Pathway

Vzorec: C₂₆H₂₈N₄O₃
MW: 444,53 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00950163
CAS číslo: 181223-80-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR