Tisk…

SL327

Dodavatel: Apollo Scientific

Synonyms: α-[amino[(4-aminophenyl)thio]methylene]-2-(trifluoromethyl)-benzeneacetonitrile, (Z)-3-amino-3-(4-aminophenyl)sulphanyl-2-[2-(trifluoromethyl)phenyl]prop-2-enenitrile, 3-Amino-3-((4-aminophenyl)thio)-2-(2-(trifluoromethyl)phenyl)acrylonitrile

BISN0193-5MG
APOSBISN0193-5MGEA 1240 CZK
APOSBISN0193-5MG
SL327
SL327

MAPK/ERK Signalling Pathway

Vzorec: C₁₆H₁₂F₃N₃S
MW: 335,4 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06411432
CAS číslo: 305350-87-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR