Tisk…

GDC0994

Dodavatel: Apollo Scientific

Synonyms: RG-7842, Ravoxertinib, (S)-1-(1-(4-Chloro-3-fluorophenyl)-2-hydroxyethyl)-4-(2-((1-methyl-1H-pyrazol-5-yl)amino)pyrimidin-4-yl)pyridin-2(1H)-one, GDC0994, GDC-0994, RG 7842, GDC 0994, RG7842, 1-[(1S)-1-(4-chloro-3-fluorophenyl)-2-hydroxyethyl]-4-[2-[(2-methylpyrazol-3-yl)amino]pyrimidin-4-yl]pyridin-2-one

BISN0188-5MG
APOSBISN0188-5MGEA 1910 CZK
APOSBISN0188-5MG
GDC0994
GDC0994

MAPK/ERK Signalling Pathway

Vzorec: C₂₁H₁₈ClFN₆O₂
MW: 440,86 g/mol
Storage Temperature: Freezer
MDL Number: MFCD28143918
CAS číslo: 1453848-26-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR