VX-11e
Dodavatel: Apollo Scientific
Synonyms:
(S)-4-(2-((2-Chloro-4-fluorophenyl)amino)-5-methylpyrimidin-4-yl)-N-(1-(3-chlorophenyl)-2-hydroxyethyl)-1H-pyrrole-2-carboxamide, VX11e, VX 11e, VX-11e, 4-[2-(2-chloro-4-fluoroanilino)-5-methylpyrimidin-4-yl]-N-[(1S)-1-(3-chlorophenyl)-2-hydroxyethyl]-1H-pyrrole-2-carboxamide
BISN0184-1MG
BISN0184-5MG
BISN0184-10MG
APOSBISN0184-1MGEA
2480
CZK
APOSBISN0184-1MG
APOSBISN0184-5MG
APOSBISN0184-10MG
VX-11e
VX-11e
MAPK/ERK Signalling Pathway
Vzorec:
C₂₄H₂₀Cl₂FN₅O₂ MW: 500,35 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD18433366 CAS číslo: 896720-20-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...