Tisk…

CGP-57380

Dodavatel: Apollo Scientific

Synonyms: N3-(4-Fluorophenyl)-1H-pyrazolo[3,4-d]pyrimidine-3,4-diamine, CGP 57380, N3-(4-Fluorophenyl)-1H-pyrazolo-[3,4-d]pyrimidine-3,4-diamine

BISN0182-1MG
APOSBISN0182-1MGEA 827 CZK
APOSBISN0182-1MG
CGP-57380
CGP-57380

MAPK/ERK Signalling Pathway

Vzorec: C₁₁H₉FN₆
MW: 244,23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03861062
CAS číslo: 522629-08-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR