Tisk…

VX-702

Dodavatel: Apollo Scientific

Synonyms: 2-(2,4-Difluorophenyl)-6-(1-(2,6-difluorophenyl)ureido)nicotinamide, 6-[(Aminocarbonyl)(2,6-difluorophenyl)amino]-2-(2,4-difluorophenyl)-3-pyridinecarboxamide

BISN0171-10MG
APOSBISN0171-10MGEA 1240 CZK
APOSBISN0171-10MG
VX-702
VX-702

MAPK/ERK Signalling Pathway

Vzorec: C₁₉H₁₂F₄N₄O₂
MW: 404,32 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11616590
CAS číslo: 745833-23-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR