Tisk…

Auraptene

Dodavatel: Apollo Scientific

Synonyms: (E)-7-((3,7-Dimethylocta-2,6-dien-1-yl)oxy)-2H-chromen-2-one, 7-[[(2E)-3,7-Dimethyl-2,6-octadien-1-yl]oxy]-2H-chromen-2-one, 7-Geranyloxycoumarin

BISN0165-250MG BISN0165-100MG
APOSBISN0165-250MGEA 3540 CZK
APOSBISN0165-250MG APOSBISN0165-100MG
Auraptene
Auraptene

MAPK/ERK Signalling Pathway

Vzorec: C₁₉H₂₂O₃
MW: 298,4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00075948
CAS číslo: 495-02-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR