Tisk…

OTS-964

Dodavatel: Apollo Scientific

Synonyms: (R)-9-(4-(1-(Dimethylamino)propan-2-yl)phenyl)-8-hydroxy-6-methylthieno[2,3-c]quinolin-4(5H)-one hydrochloride, (R)-9-(4-(1-(dimethylamino)propan-2-yl)phenyl)-8-hydroxythieno[2,3-c]quinolin-4(5H)-one, OTS964 HCl

BISN0160-2MG
APOSBISN0160-2MGEA 2200 CZK
APOSBISN0160-2MG
OTS-964
OTS-964

MAPK/ERK Signalling Pathway

Vzorec: C₂₂H₂₂N₂O₂S
MW: 378,49 g/mol
Storage Temperature: Freezer
MDL Number: MFCD28386220
CAS číslo: 1338545-07-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR