Tisk…

SCH772984

Dodavatel: Apollo Scientific

Synonyms: SCH-772984, (R)-1-(2-Oxo-2-(4-(4-(pyrimidin-2-yl)phenyl)piperazin-1-yl)ethyl)-N-(3-(pyridin-4-yl)-1H-indazol-5-yl)pyrrolidine-3-carboxamide, (3R)-1-[2-oxo-2-[4-(4-pyrimidin-2-ylphenyl)piperazin-1-yl]ethyl]-N-(3-pyridin-4-yl-1H-indazol-5-yl)pyrrolidine-3-carboxamide, SCH 772984, SCH772984

BISN0139-2MG BISN0139-5MG BISN0139-10MG
APOSBISN0139-2MGEA 3540 CZK
APOSBISN0139-2MG APOSBISN0139-5MG APOSBISN0139-10MG
SCH772984
SCH772984

SCH772984 is a specific inhibitor of ERK12 with IC50 of 4 nM and 1 nM respectively.

Vzorec: C₃₃H₃₃N₉O₂
MW: 587,67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28144700
CAS číslo: 942183-80-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR