Tisk…

Oxaliplatin

Dodavatel: Apollo Scientific

BISN0119-50MG BISN0119-100MG BISN0119-250MG
APOSBISN0119-50MGEA 3110 CZK
APOSBISN0119-50MG APOSBISN0119-100MG APOSBISN0119-250MG
Oxaliplatin
Oxaliplatin

Oxaliplatin inhibits DNA synthesis by conforming DNA adducts.

Vzorec: C₈H₁₄N₂O₄Pt
Storage Temperature: Ambient
MDL Number: MFCD00866327
CAS číslo: 61825-94-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR