Chloroquine diphosphate salt
Dodavatel: Apollo Scientific
Synonyms:
N4-(7-Chloroquinolin-4-yl)-N1,N1-diethylpentane-1,4-diamine bis(phosphate), N4-(7-Chloro-4-quinolyl)-N1,N1-diethyl-1,4-pentanediamine Diphosphate, 7-Chloro-4-[4-(diethylamino)-1-methylbutylamino]quinoline Diphosphate, Chloroquine phosphate
BISN0106-25MG
BISN0106-100MG
APOSBISN0106-25MGEA
2280
CZK
APOSBISN0106-25MG
APOSBISN0106-100MG
Chloroquine diphosphate salt
Chloroquine diphosphate salt
Chloroquine Diphosphate is an inhibitor of autophagylysosome and Toll-like receptor TLR signalling also acting as an ATM acitivator its an antimalarial drug.
Vzorec:
C₁₈H₂₆ClN₃ MW: 515.86 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00069852 CAS číslo: 50-63-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...