Pravastatin sodium
Dodavatel: Apollo Scientific
Synonyms:
(3R,5R)-3,5-Dihydroxy-7-((1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-(((S)-2-methylbutanoyl)oxy)-1,2,6,7,8,8a-hexahydronaphthalen-1-yl)heptanoic acid, sodium salt
BISN0103-100MG
APOSBISN0103-100MGEA
0
CZK
APOSBISN0103-100MG
Pravastatin sodium
Pravastatin sodium
Pravastatin Sodium is a competitive HMG-CoA reductase inhibitor with IC50 value of 56 M.
Vzorec:
C₂₃H₃₅NaO₇ MW: 446,52 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00887601 CAS číslo: 81131-70-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...