Tisk…

EX 527

Dodavatel: Apollo Scientific

Synonyms: 6-Chloro-2,3,4,9-tetrahydro-1H-carbazole-1-carboxamide, EX-527, Selisistat

BISN0099-25MG
APOSBISN0099-25MGEA 0 CZK
APOSBISN0099-25MG
EX 527
EX 527

EX-527 is a potent and selective SIRT1 inhibitor with IC50 of 38 nM, exhibits 200-fold selectivity against SIRT2 and SIRT3.

Vzorec: C₁₃H₁₃ClN₂O
MW: 248,71 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03009471
CAS číslo: 49843-98-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR