Tisk…

SC-79

Dodavatel: Apollo Scientific

Synonyms: Ethyl 2-amino-6-chloro-4-(1-cyano-2-ethoxy-2-oxoethyl)-4H-chromene-3-carboxylate, 2-Amino-6-chloro-α-cyano-3-(ethoxyca­rbonyl)-4H-1-benzopyran-4-acetic acid ethyl ester

BISN0098-1MG BISN0098-5MG BISN0098-25MG
APOSBISN0098-1MGEA 2280 CZK
APOSBISN0098-1MG APOSBISN0098-5MG APOSBISN0098-25MG
SC-79
SC-79

SC79 is a unique specific Akt activator that may be used to enhance Akt activity in various physiological and pathological conditions.

Vzorec: C₁₇H₁₇ClN₂O₅
MW: 364,78 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02681303
CAS číslo: 305834-79-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR