Tisk…

GLX-351322 98%

Dodavatel: Apollo Scientific

BISN0056-5MG BISN0056-25MG
APOSBISN0056-5MGEA 2110 CZK
APOSBISN0056-5MG APOSBISN0056-25MG
GLX-351322 98%
GLX-351322

GLX351322 is an inhibitor of NADPH oxidase 4, inhibits hydrogen peroxide production from NOX4-overexpressing cells with an IC50 of 5 μ M.

Vzorec: C₂₁H₂₅N₃O₅S
Storage Temperature: Ambient
MDL Number: MFCD05123600
CAS číslo: 835598-94-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR