Tisk…

Edicotinib 99%

Dodavatel: Apollo Scientific

Synonyms: 5-Cyano-N-(2-(4,4-dimethylcyclohex-1-en-1-yl)-6-(2,2,6,6-tetramethyltetrahydro-2H-pyran-4-yl)pyridin-3-yl)-1H-imidazole-2-carboxamide, PRV 6527, JNJ 40346527, 4-cyano-N-[2-(4,4-dimethylcyclohex-1-en-1-yl)-6-(2,2,6,6-tetramethyloxan-4-yl)pyridin-3-yl]-1H-imidazole-2-carboxamide

BISN0042-50MG
APOSBISN0042-50MGEA 7080 CZK
APOSBISN0042-50MG
Edicotinib 99%
Edicotinib

JNJ-40346527 is an inhibitor of colony-stimulating factor-1 receptor (CSF1R) with potential antineoplastic activity.

Vzorec: C₂₇H₃₅N₅O₂
MW: 461,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30489233
CAS číslo: 1142363-52-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR