UNC0224 98%
Dodavatel: Apollo Scientific
Synonyms:
7-(3-(Dimethylamino)propoxy)-6-methoxy-2-(4-methyl-1,4-diazepan-1-yl)-N-(1-methylpiperidin-4-yl)quinazolin-4-amine
BISN0023-5MG
BISN0023-10MG
BISN0023-25MG
APOSBISN0023-5MGEA
4570
CZK
APOSBISN0023-5MG
APOSBISN0023-10MG
APOSBISN0023-25MG
UNC0224 98%
UNC0224
UNC0224 is a selective inhibitor of G9a with IC50 of 15 nM.
Vzorec:
C₂₆H₄₃N₇O₂ MW: 485,67 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18251564 CAS číslo: 1197196-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...