Tisk…
SEW-2871 98%
Dodavatel: Apollo Scientific
Synonyms: 5-(4-Phenyl-5-(trifluoromethyl)thiophen-2-yl)-3-(3-(trifluoromethyl)phenyl)-1,2,4-oxadiazole, 5-[4-Phenyl-5-(trifluoromethyl)thiophen-2-yl]-3-[3-(trifluoromethyl)phenyl]-1,2,4-oxadiazole, SEW2871
BISN0002-5MG BISN0002-10MG BISN0002-25MG
APOSBISN0002-5MGEA 3440 CZK
APOSBISN0002-5MG APOSBISN0002-10MG APOSBISN0002-25MG
SEW-2871 98%
SEW-2871
SEW2871 is a potent and selective sphingosine-1-phosphate 1 (S1P1) receptor agonist. Activates S1P1 receptor with an EC50 of 13 nM, but does not activate S1P2, S1P3, S1P4 or S1P5 receptors at concentrations up to 10 μM. Cell-permeable and active in vivo.
Vzorec: C₂₀H₁₀F₆N₂OS
MW: 440.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00096600
CAS číslo: 256414-75-2