Sinapic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3,5-Dimethoxy-4-hydroxycinnamic acid, 3',5'-Dimethoxy-4'-hydroxycinnamic acid, 4-Hydroxy-3,5-dimethoxycinnamic acid
BIS8112-5G
BIS8112-25G
BIS8112-100G
APOSBIS8112-5GEA
622
CZK
APOSBIS8112-5G
APOSBIS8112-25G
APOSBIS8112-100G
Sinapic acid 98%
Sinapic acid
- Protein
Vzorec:
C₁₁H₁₂O₅ MW: 224,21 g/mol Bod tání: 195…202 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004401 CAS číslo: 530-59-6 EINECS: 208-487-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...