Spermine tetrahydrochloride 99%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
N1,N1'-(Butane-1,4-diyl)bis(propane-1,3-diamine) tetrahydrochloride
BIS5943-10G
BIS5943-25G
BIS5943-5G
APOSBIS5943-10GEA
4770
CZK
APOSBIS5943-10G
APOSBIS5943-25G
APOSBIS5943-5G
Spermine tetrahydrochloride 99%
Spermine tetrahydrochloride
For precipitation of DNA from aqueous buffers
Vzorec:
C₁₀H₃₀Cl₄N₄ MW: 348,19 g/mol Bod tání: 310…311 °C Storage Temperature: Freezer |
MDL Number:
MFCD00012914 CAS číslo: 306-67-2 EINECS: 206-189-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...