Syringaldazine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-[[(3,5-dimethoxy-4-oxo-1-cyclohexa-2,5-dienylidene)methylhydrazo]methylidene]-2,6-dimethoxy-1-cyclohexa-2,5-dienone, 4-Hydroxy-3,5-dimethoxybenzaldehyde azine, 3,5-Dimethoxy-4-hydroxybenzalazine, 4,4'-(Hydrazine-1,2-diylidenebis(methanylylidene))bis(2,6-dimethoxyphenol)
BIS1000-10G
BIS1000-25G
BIS1000-50G
APOSBIS1000-10GEA
3940
CZK
APOSBIS1000-10G
APOSBIS1000-25G
APOSBIS1000-50G
Syringaldazine
Syringaldazine
Vzorec:
C₁₈H₂₀N₂O₆ MW: 360,36 g/mol Bod tání: 209…210 °C Storage Temperature: Freezer |
MDL Number:
MFCD00008363 CAS číslo: 14414-32-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...