Tisk…

Sarcosine hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(Methylamino)acetic acid hydrochloride

BIS0907-100G BIS0907-25G BIS0907-5G
APOSBIS0907-100GEA 3110 CZK
APOSBIS0907-100G APOSBIS0907-25G APOSBIS0907-5G
Sarcosine hydrochloride
Sarcosine hydrochloride
Vzorec: C₃H₈ClNO₂
MW: 125,55 g/mol
Bod tání: 173…175 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00012609
CAS číslo: 637-96-7
EINECS: 211-310-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR