Rapamycin (from Streptomyces hygroscopicus)
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(3S,6R,7E,9R,10R,12R,14S,15E,17E,19E,21S,23S,26R,27R,34aS)-9,10,12,13,14,21,22,23,24,25,26,27,32,33,34,34a-Hexadecahydro-9,27-dihydroxy-3-[(1R)-2-[(1S,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethyl]-10,21-dimethoxy-6,8,12,14,20,26-hexamethyl-23,27-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine-1,5,11,28,29(4H,6H,31H)-pentone
Macrocyclic triene antibiotic.
Vzorec:
C₅₁H₇₉NO₁₃ MW: 914,19 g/mol Bod tání: 183…185 °C Storage Temperature: Freezer |
MDL Number:
MFCD00867594 CAS číslo: 53123-88-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...