Tisk…

t-Boc-N-amido-PEG4-acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2,2-Dimethyl-4-oxo-3,8,11,14,17-pentaoxa-5-azaicosan-20-oic acid

BIPG1802-500MG BIPG1802-100MG BIPG1802-1G
APOSBIPG1802-500MGEA 7870 CZK
APOSBIPG1802-500MG APOSBIPG1802-100MG APOSBIPG1802-1G
t-Boc-N-amido-PEG4-acid
t-Boc-N-amido-PEG4-acid
Vzorec: C₁₆H₃₁NO₈
MW: 365,42 g/mol
Storage Temperature: Refrigerator
CAS číslo: 756525-91-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR