Methyl-PEG12-carboxylic acid
Dodavatel: Apollo Scientific
Synonyms:
2,5,8,11,14,17,20,23,26,29,32,35-Dodecaoxaoctatriacontan-38-oic acid, m-PEG12-acid
BIPG1551-100MG
BIPG1551-50MG
APOSBIPG1551-100MGEA
13460
CZK
APOSBIPG1551-100MG
APOSBIPG1551-50MG
Methyl-PEG12-carboxylic acid
Methyl-PEG12-carboxylic acid
Vzorec:
C₂₆H₅₂O₁₄ MW: 588,69 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08274611 CAS číslo: 2135793-73-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...